Learn More
Corning™ Cell Culture Buffers: Dulbecco's Phosphate-Buffered Salt Solution 1X
BIOPHARMA
Product Code. 15333631
Click to view available options
Quantity:
6 x 500 mL
6 x 1000 mL
Physical Form:
Liquid
Includes:
Calcium and Magnesium
No Calcium, No Magnesium
Unit Size:
Pack of 6
Product Code. 15333631
Brand: Corning™ 21030CM
With low endotoxin content.
Dulbecco's Phosphate Buffered Saline (DPBS) is a buffer solution which can be used to maintain cell culture media in the physiological pH range of 7.0 to 7.6. It can serve as an irrigating, transporting or diluting fluid while maintaining cell tonicity and viability for a limited period of time. This formulation contains calcium and magnesium.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material DPBS
Concentration 1 X
pH 6.9 to 7.1
Physical Form Liquid
Quantity 6 x 1000 mL
Formulation Calcium Chloride 0.10g/L, Potassium Chloride 0.2g/L, Potassium Dihydrogen Phosphate 0.2g/L, Magnesium Chloride Hexahydrate 0.1g/L, Sodium Chloride 8g/L, Disodium Phosphate Heptahydrate 2.1716g/L
Includes Calcium and Magnesium