missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated YTHDF2 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
129.00€ - 395.00€
Specifications
| Antigen | YTHDF2 |
|---|---|
| Concentration | 1000 μg/mL |
| Applications | Not Available |
| Classification | Polyclonal |
| Conjugate | CoraLite Plus 488 |
Description
CoraLite™ Plus 488-conjugated YTHDF2 Rabbit Polyclonal Antibody, Proteintechsh ows reactivity in Human, Mouse and Rat samples.Specifications
| YTHDF2 | |
| Not Available | |
| CoraLite Plus 488 | |
| Rabbit | |
| Human, Mouse, Rat | |
| HGRG8; NY-REN-2 | |
| IgG | |
| Antigen affinity purification | |
| Conjugated Antibody |
| 1000 μg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 51441 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title