missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated VEGFA Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66828-100UL
This item is not returnable.
View return policy
Description
The VEGFA antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human VEGFA. This antibody recognizes human antigen. The VEGFA antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™ plus 488.Specifications
| VEGFA | |
| Monoclonal | |
| 1000 μg/mL | |
| MGC70609; MVCD1; VEGF; VEGF-A; VPF | |
| Recombinant Protein | |
| 100 μL | |
| Primary | |
| Human | |
| 43 kDa | |
| Liquid |
| Flow Cytometry (Intracellular Staining) | |
| 2E2H9 | |
| CoraLite Plus 488 | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 7422 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Conjugated Antibody | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction