missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated Syntaxin 1A / Syntaxin 1B Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66437-20UL
This item is not returnable.
View return policy
Description
The Syntaxin 1A / Syntaxin 1B antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Syntaxin 1A / Syntaxin 1B. This antibody recognizes human, mouse, rat antigen. The Syntaxin 1A / Syntaxin 1B antibody has been validated for the following applications: IF-P analysis. This antibody is coralite™ plus 488.Specifications
| Syntaxin 1A / Syntaxin 1B | |
| Monoclonal | |
| 1000 μg/mL | |
| STX1B1; STX1B2 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| 33 kDa | |
| Liquid |
| Immunofluorescence | |
| 2D3B5 | |
| CoraLite Plus 488 | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 112755 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Conjugated Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction