missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated PPARA Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
129.00€ - 349.00€
Specifications
| Antigen | PPARA |
|---|---|
| Clone | 1G1E10 |
| Concentration | 1000 μg/mL |
| Applications | Not Available |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30236679
|
Proteintech
CL488-66826-100UL |
100 μL |
361.81€ 349.00€ / 100µL Save 12.81€ 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30237519
|
Proteintech
CL488-66826-20UL |
20 μL |
133.74€ 129.00€ / 20µL Save 4.74€ 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CoraLite™ Plus 488-conjugated PPARA Mouse Monoclonal Antibody, Proteintech shows reactivity in Human, Rat samples.Specifications
| PPARA | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 5465 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 1G1E10 | |
| Not Available | |
| CoraLite Plus 488 | |
| Mouse | |
| Human, Rat | |
| MGC2237; MGC2452; NR1C1; PPAR; hPPAR | |
| IgG1 | |
| Protein G purification | |
| Conjugated Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title