missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated Phospho-Histone H3 (Ser10) Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
129.00€ - 349.00€
Specifications
| Antigen | Histone H3 (Ser10) |
|---|---|
| Clone | 4C7G2 |
| Concentration | 1000 μg/mL |
| Applications | Flow Cytometry (Intracellular Staining) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30234478
|
Proteintech
CL488-66863-100UL |
100 μL |
450.00€ 349.00€ / 100µL Save 101.00€ 22% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30236932
|
Proteintech
CL488-66863-20UL |
20 μL |
140.00€ 129.00€ / 20µL Save 11.00€ 7% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Phospho-Histone H3 (Ser10) antibody from Proteintech is a mouse monoclonal antibody generated with peptide of human Phospho-Histone H3 (Ser10). This antibody recognizes human, mouse, rat antigen. The Phospho-Histone H3 (Ser10) antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™ plus 488.Specifications
| Histone H3 (Ser10) | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 8350 | |
| IgG1 | |
| Protein G purification | |
| Conjugated Antibody |
| 4C7G2 | |
| Flow Cytometry (Intracellular Staining) | |
| CoraLite Plus 488 | |
| Mouse | |
| Human, Mouse, Rat | |
| Peptide | |
| Primary | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title