missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated PARK2/Parkin Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
129.00€ - 395.00€
Specifications
| Antigen | PARK2/Parkin |
|---|---|
| Concentration | 1000 μg/mL |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | CoraLite Plus 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30236191
|
Proteintech
CL488-14060-100UL |
100 μL |
450.00€ 395.00€ / 100µL Save 55.00€ 12% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30234652
|
Proteintech
CL488-14060-20UL |
20 μL |
140.00€ 129.00€ / 20µL Save 11.00€ 7% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The PARK2/Parkin antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PARK2/Parkin. This antibody recognizes human, mouse, rat antigen. The PARK2/Parkin antibody has been validated for the following applications: IF-P analysis. This antibody is coralite™ plus 488.Specifications
| PARK2/Parkin | |
| Immunofluorescence | |
| CoraLite Plus 488 | |
| Rabbit | |
| Human, Mouse, Rat | |
| AR-JP; LPRS2; PDJ; PRKN | |
| IgG | |
| Antigen affinity purification | |
| Conjugated Antibody |
| 1000 μg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 5071 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title