missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated Myosin Light Chain 2/MLC-2V Rabbit Polyclonal Antibody, Proteintech
Shop All Proteintech ProductsDescription
The Myosin Light Chain 2/MLC-2V antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Myosin Light Chain 2/MLC-2V. This antibody recognizes human, mouse, rat, zebrafish antigen. The Myosin Light Chain 2/MLC-2V antibody has been validated for the following applications: IF-P analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
| Antigen | Myosin Light Chain 2/MLC-2V |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Concentration | 1000 μg/mL |
| Conjugate | CoraLite Plus 488 |
| Gene Symbols | CMH10; DKFZp779C0562; MLC2 |
| Host Species | Rabbit |
| Immunogen | Recombinant Protein |
| Purification Method | Antigen affinity purification |
| Quantity | 20 μL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?