missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated EIF2S1 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
129.00€ - 395.00€
Specifications
| Antigen | EIF2S1 |
|---|---|
| Concentration | 1000 μg/mL |
| Applications | Flow Cytometry (Intracellular Staining) |
| Classification | Polyclonal |
| Conjugate | CoraLite Plus 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30234218
|
Proteintech
CL488-11170-20UL |
20 μL |
140.00€ 129.00€ / 20µL Save 11.00€ 7% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30233259
|
Proteintech
CL488-11170-100UL |
100 μL |
450.00€ 395.00€ / 100µL Save 55.00€ 12% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The EIF2S1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human EIF2S1. This antibody recognizes human, mouse, rat antigen. The EIF2S1 antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™ plus 488.Specifications
| EIF2S1 | |
| Flow Cytometry (Intracellular Staining) | |
| CoraLite Plus 488 | |
| Rabbit | |
| Human, Mouse, Rat | |
| EIF-2; EIF-2A; EIF-2alpha; EIF2; EIF2A | |
| IgG | |
| Antigen affinity purification | |
| Conjugated Antibody |
| 1000 μg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 1965 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title