missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated CD97 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
129.00€ - 349.00€
Specifications
| Antigen | CD97 |
|---|---|
| Clone | 3F10C2 |
| Concentration | 1000 μg/mL |
| Applications | Not Available |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30237676
|
Proteintech
CL488-66972-20UL |
20 μL |
133.74€ 129.00€ / 20µL Save 4.74€ 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30237172
|
Proteintech
CL488-66972-100UL |
100 μL |
361.81€ 349.00€ / 100µL Save 12.81€ 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descripción
CoraLite™ Plus 488-conjugated CD97 Mouse Monoclonal Antibody, Proteintech shows reactivity in Human, Mouse samples.Especificaciones
| CD97 | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 976 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 3F10C2 | |
| Not Available | |
| CoraLite Plus 488 | |
| Mouse | |
| Human, Mouse | |
| TM7LN1 | |
| IgG2a | |
| Protein A purification | |
| Conjugated Antibody |
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto