missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated AR Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66747-20UL
This item is not returnable.
View return policy
Description
The AR antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human AR. This antibody recognizes human antigen. The AR antibody has been validated for the following applications: IF/ICC analysis. This antibody is coralite™ plus 488.Specifications
| AR | |
| Monoclonal | |
| 1000 μg/mL | |
| AIS; DHTR; HUMARA; HYSP1; KD; NR3C4; SBMA; SMAX1; TFM | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human | |
| 110-120 kDa | |
| Liquid |
| Immunocytochemistry/Immunofluorescence | |
| 1F7C12 | |
| CoraLite Plus 488 | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 367 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Conjugated Antibody | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction