missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated APBB1 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
129.00€ - 349.00€
Specifications
| Antigen | APBB1 |
|---|---|
| Clone | 1B9G10 |
| Concentration | 1000 μg/mL |
| Applications | Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30233144
|
Proteintech
CL488-67077-100UL |
100 μL |
450.00€ 349.00€ / 100µL Save 101.00€ 22% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30236377
|
Proteintech
CL488-67077-20UL |
20 μL |
140.00€ 129.00€ / 20µL Save 11.00€ 7% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The APBB1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human APBB1. This antibody recognizes human, mouse, rat, pig antigen. The APBB1 antibody has been validated for the following applications: IF-P analysis. This antibody is coralite™ plus 488.Specifications
| APBB1 | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 322 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 1B9G10 | |
| Immunofluorescence | |
| CoraLite Plus 488 | |
| Mouse | |
| Human, Mouse, Rat, Pig | |
| FE65; MGC:9072; RIR | |
| IgG1 | |
| Protein G purification | |
| Conjugated Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title