missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated Annexin A2 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66035-100UL
This item is not returnable.
View return policy
Description
The Annexin A2 antibody from Proteintech is a mouse monoclonal antibody generated with fusion protein of human Annexin A2. This antibody recognizes human antigen. The Annexin A2 antibody has been validated for the following applications: IF/ICC analysis. This antibody is coralite™ plus 488.Specifications
| Annexin A2 | |
| Monoclonal | |
| 1000 μg/mL | |
| ANX2; ANX2L4; CAL1H; LIP2; LPC2; LPC2D; P36; PAP-IV | |
| Fusion Protein | |
| 100 μL | |
| Primary | |
| Human | |
| 39 kDa | |
| Liquid |
| Immunocytochemistry/Immunofluorescence | |
| 1C1E12 | |
| CoraLite Plus 488 | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 302 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Conjugated Antibody | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction