missing translation for 'onlineSavingsMsg'
Learn More

CoraLite™ Plus 488-conjugated ACLY Mouse Monoclonal Antibody, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech CL488-67166-100UL

 View more versions of this product

Product Code. 30234253

  • ONLINE EXCLUSIVE PRICE
    450.00€
    349.00€ / 100µL
    Save 101.00€
    22% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The ACLY antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ACLY. This antibody recognizes human, mouse, rat antigen. The ACLY antibody has been validated for the following applications: IF/ICC analysis. This antibody is coralite™ plus 488.
Specifications

Specifications

ACLY
Monoclonal
1000 μg/mL
ACL; ATPCL; CLATP
Recombinant Protein
100 μL
Primary
Human, Mouse, Rat
120 kDa
Liquid
Immunocytochemistry/Immunofluorescence
1E10D5
CoraLite Plus 488
Mouse
Caprylic acid/ammonium sulfate precipitation
RUO
47
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Conjugated Antibody
IgM
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.