missing translation for 'onlineSavingsMsg'
Learn More

CoraLite™ Plus 488-conjugated Acetyl-Tubulin (Lys40) Mouse Monoclonal Antibody, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech CL488-66200-20UL

Product Code. 30233923

  • ONLINE EXCLUSIVE PRICE
    140.00€
    129.00€ / 20µL
    Save 11.00€
    7% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The Acetyl-Tubulin (Lys40) antibody from Proteintech is a mouse monoclonal antibody generated with peptide of human Acetyl-Tubulin (Lys40). This antibody recognizes human, mouse, rat, pig, canine antigen. The Acetyl-Tubulin (Lys40) antibody has been validated for the following applications: IF/ICC, IF-Fro analysis. This antibody is coralite™ plus 488.
Specifications

Specifications

Acetyl-Tubulin (Lys40)
Monoclonal
2000 μg/mL
B-ALPHA-1; FLJ25113; LIS3; TUBA3
Peptide
20 μL
Primary
Human, Mouse, Rat, Pig, Canine
Conjugated Antibody
IgG1
Immunocytochemistry/Immunofluorescence, Immunofluorescence
7E5H8
CoraLite Plus 488
Mouse
Protein G purification
RUO
7846
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.