missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COPB2 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33309-100ul
This item is not returnable.
View return policy
Description
COPB2 Monoclonal antibody specifically detects COPB2 in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| COPB2 | |
| Monoclonal | |
| Western Blot 1:3000 - 1:20000, ELISA Recommended starting concentration is 1 μg/mL | |
| beta prime subunit, Beta'-coat protein, Beta'-COP, betaprime-COP, coatomer protein complex, subunit beta 2 (beta prime), coatomer subunit beta', p102 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 657-906 of human COPB2 (NP_004757.1).,, Sequence:, AYQLAVEAESEQKWKQLAELAISKCQFGLAQECLHHAQDYGGLLLLATASGNANMVNKLAEGAERDGKNNVAFMSYFLQGKVDACLELLIRTGRLPEAAFLARTYLPSQVSRVVKLWRENLSKVNQKAAESLADPTEYENLFPGLKEAFVVEEWVKETHADLWPAKQYPLVTPNEERNVMEEGKDFQPSRSTAQQELDGKPASPTPVIVASHTANKEEKSLLELEVDLDNLELEDIDTTDINLDEDILDD | |
| 100 μL | |
| Golgi Apparatus Markers, Membrane Trafficking and Chaperones, Neuroscience | |
| 9276 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction