missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COP9 signalosome complex subunit 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-58587-25ul
This item is not returnable.
View return policy
Description
COP9 signalosome complex subunit 2 Polyclonal specifically detects COP9 signalosome complex subunit 2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| COP9 signalosome complex subunit 2 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 1-4 μg/mL | |
| ALIEN, Alien homolog, COP9 constitutive photomorphogenic homolog subunit 2 (Arabidopsis), COP9 signalosome complex subunit 2, CSN2JAB1-containing signalosome subunit 2, SGN2TR-interacting protein 15, Signalosome subunit 2, thyroid receptor interacting protein 15, Thyroid receptor-interacting protein 15, TRIP15TRIP-15 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol with 0.02% Sodium Azide | |
| COPS2 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:ISTSKQNSDFLCQMDLLQEFYETTLEALKDAKNDRLWFKTNTKLGKLYLEREEYGKLQKILRQLHQSCQTDDGEDDLKKGTQLLEIYALEIQMYTAQKNNKKLKALYEQ | |
| 25 μL | |
| Cell Cycle and Replication, Neuronal Cell Markers, Neurotransmission, Signal Transduction | |
| 9318 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction