Learn More
Thermo Scientific™ Consumables and Services Kits for Orion™ 2110XP Ammonia Analyzer
Product Code. p-4564069
Click to view available options
Type:
Ammonia Calibration Kit
Ammonia Consumables Kit
Unit Size:
Each
Product Code. 17128988
Brand: Thermo Scientific™ 211050XP
Keep needed supplies and service items in stock to avoid lost productivity with Thermo Scientific™ Consumables and Services Kits for Orion™ 2110XP Ammonia Analyzer.
Keep needed supplies and service items in stock to avoid lost productivity with Thermo Scientific™ Consumables and Services Kits for the Thermo Scientific™ Orion™ 2110XP Ammonia Analyzer.
TRUSTED_SUSTAINABILITY
Specifications
Accuracy (mV) ±(0.5 mV + 0.1%)
Accuracy (Temperature) ±0.5 °C
Description Ammonia Consumables Kit for one year operation
Temperature (Metric) Operating -10°C to +120°C
Type Ammonia Consumables Kit
Display Type 211050XP
Flow Rate 40 mL/min.
Includes 2 x 2L acetic acid reagent (181030), 2 x 4 ft. diffusion tubing assemblies with O-rings (150060), reagent bottle adapter (2110RBA), ammonia calibration standards kit (181040), ammonia electrode (100047), reference electrode (210056), 5 x 2 oz. reference electrode filling solution (181073), O-ring kit (2110OK), restriction tube assembly (2100RT), 4 inlet filters (181120) and box of pipette tips (204846-001)
Inlet Pressure 8 to 100 psig
Inputs 85-132; 170-264 VAC
Outputs Alarm: 3; Relay: 3; Analog: 2; Selections: 0-20mA; 4-20 mA
Range (mV) ±1999.9 m
Relay Details Max Load: 250VAC; 30VDC; 5A
Resolution (mV) 0.1 mV
Resolution (Temperature) 0.1 °C
Response Time 211050XP
Sample Inlet 0.25 NPTF Tube Fitting in.
Sample Temperature 5 to 45 °C