Learn More
Connexin 43 Rabbit, anti-Human, CoraLite Plus 488, Clone: 3O6, Proteintech
Rabbit Recombinant Monoclonal Antibody
Brand: Proteintech CL488-80543-20UL
Description
The Connexin 43 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human Connexin 43. This antibody recognizes human, mouse, rat, pig antigen. The Connexin 43 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| Connexin 43 | |
| 3O6 | |
| CoraLite Plus 488 | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat, Pig | |
| 150 kDa | |
| IgG |
| Immunofluorescence | |
| 1000 μg/mL | |
| Connexin 43, CX43, DFNB38, Gap junction alpha 1 protein, GJA1, GJAL, ODDD | |
| 20 μL | |
| 2697 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |