missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Connexin 40/GJA5 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
193.00€ - 463.00€
Specifications
| Antigen | Connexin 40/GJA5 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18604282
|
Novus Biologicals
NBP2-92491-0.02ml |
0.02 mL |
193.00€
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18692282
|
Novus Biologicals
NBP2-92491-0.1ml |
0.1 mL |
463.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Connexin 40/GJA5 Polyclonal antibody specifically detects Connexin 40/GJA5 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| Connexin 40/GJA5 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cardiovascular Biology, Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| PBS with 50% glycerol, pH7.3. | |
| 2702 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| alpha 5, 40kD (connexin 40), connexin-40, Cx40, gap junction alpha-5 protein, gap junction protein, alpha 5, 40kDa, gap junction protein, alpha 5, 40kDa (connexin 40), MGC11185 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 229-358 of human GJA5 (NP_859054.1). LGWKKIRQRFVKPRQHMAKCQLSGPSVGIVQSCTPPPDFNQCLENGPGGKFFNPFSNNMASQQNTDNLVTEQVRGQEQTPGEGFIQVRYGQKPEVPNGVSPGHRLPHGYHSDKRRLSKASSKARSDDLSV | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title