missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Reagecon offers high quality Conductivity Standards that support accurate conductivity measurements and can be used for sensor calibration, quality control, method validation, and instrument qualification.
- The standards are aqueous-based, minimising errors caused by matrix mismatch.
- Directly traceable to NIST reference materials, meeting the specification requirements of all major Pharmacopoeias.
- Provided with a comprehensive Certificate of Analysis.
- A temperature conductivity table can be accessed via the product label and clearly displayed lot numbers and expiry dates for ease of use.
- A low temperature coefficient reduces temperature related errors, allowing reliable measurements without strict temperature control and supporting field use.
- High accuracy allows the standards function effectively as both calibrators and controls for demanding industrial applications.
- Manufactured to stringent specifications, ensuring extended shelf life and stability.
Specifications
Specifications
| Chemical Name or Material | Conductivity Standard |
| Packaging | HDPE Bottle |
| Conductivity | 1413 μS/cm ±1.0% |
| Type | Certified Reference Material |
| Molecular Weight (g/mol) | Unknown |
| Quantity | 500 mL |
| Grade | Reference Material |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?