Learn More
84 μS/cm Conductivity Standards, Hanna Instruments™
Product Code. 11707064
Click to view available options
Quantity:
500 mL
Unit Size:
Each
Product Code. 11707064
Brand: Hanna Instruments HI7033LC
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Premium quality 84 μS/cm conductivity standard that is prepared against a NIST traceable potassium chloride solution.
  • Calibrated with potassium chloride NIST 2202 SRM (Standard Reference Material) in deionized water for analytical use in accordance with ISO 3696/BS3978
  • Temperature chart of the actual μS/cm value at various temperatures printed on each bottle
  • Premium quality 84 μS/cm conductivity solution standardized with a conductivity meter
  • Additional equipment used in preparation of the solutions include certified weight-checked balances, Class A glassware and certified thermometers
  • Air tight bottle with tamper-proof seal to ensure quality
Specifications
Packaging Bottles
Quantity 500 mL