Learn More
Basic Line Conductivity Standard, 1413μS/cm, Hamilton™
Product Code. 10607735
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10607735 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10607735 Supplier Hamilton Supplier No. 238986
  • ONLINE EXCLUSIVE PRICE
    94.49€
    69.11€ / 500mL
    Save 25.38€
    26% Off
 Restricted Item
Estimated Shipment: 20-11-2026
Provides unprecedented stability. Hamilton™ Basic Line Conductivity Standard, 1413μS/cm comes with a certificate with calibration document form DFM.
  • Remains stable for up to 3 years
  • Expiration date shown on every bottle
  • Bottles are permitted to stay open for a total of 60 minutes
Specifications
Packaging Calpack bottle
Quantity 500 mL