Learn More
Conductivity Solution, 100μS.cm-1, Fisher Chemical™
BIOPHARMA
Product Code. 12974485 Shop All Fisher Chemical Products
Click to view available options
Quantity:
500 mL
1000 mL
Unit Size:
1000mL
500mL
Product Code. 12974485
Brand: Fisher Chemical CTE 005 500
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Specifications
Conductivity 100μS.cm-1
Physical Form Liquid
Quantity 500 mL