missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Reagecon™ Conductivity Resistivity Standard Solution, 26.6μS/cm
Brand: Reagecon™ EP266
Additional Details : Weight : 0.80000kg
| Quantity | 500mL |
|---|
Specifications
| Liquid | |
| Not miscible or difficult to mix | |
| Colorless | |
| 26.6μS/cm |
| Characteristic | |
| 1.00015g/cm3 at 20°C | |
| 500mL |