missing translation for 'onlineSavingsMsg'
Learn More

Abcam Concanamycin C, H+-ATPase Inhibitor, Solid, 25 μg

Product Code. 32173639
Change view
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32173639 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32173639 Supplier Abcam Supplier No. ab14422925ug
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Potent and selective vacuolar - type H+ - ATPase inhibitor.

  • Molecular Weight: 823.1 Da
  • Purity: >98%
  • Action: Potent, selective vacuolar-type H+-ATPase inhibitor
  • Classification: Macrolide antibiotic agent
  • Analogue: Concanamycin A (ab144227) analog
  • Biological Effects: Shows antiproliferative and potent antifungal effects
  • Form: Solid

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Buffer 100% Concanamycin C
Content And Storage Store at -20°C.
Form Solid
Shipping Condition Ambient
Substrate Properties Chemical Substrate
Target Molecule H+-ATPase
Quantity 25 μg
For Use With (Application) H+-ATPase inhibition, antiproliferative research, antifungal research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.