missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Molecular Weight: 852.1 Da
- Purity: >98%
- Physical Form: Solid
- Mechanism of Action: Potent, selective vacuolar type H+-ATPase inhibitor
- Biological Activity: Antibiotic agent; induces EGF-dependent apoptosis; shows potent antiviral effects (IC50 = 6 nM, HSV-1 replication)
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Content And Storage | Store at -20°C. |
| Form | Solid |
| Shipping Condition | Ambient |
| Target Molecule | H+-ATPase |
| Formulation | 100% Concanamycin B |
| Quantity | 500 μg |
| For Use With (Application) | H+-ATPase inhibition, Apoptosis induction, Antiviral research |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?