missing translation for 'onlineSavingsMsg'
Learn More

Abcam Complex III Immunocapture Kit, 250 μg

Product Code. 32108690 Shop All Abcam Products
Change view
Click to view available options
Quantity:
250 μg
Unit Size:
250µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32108690 250 μg 250µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32108690 Supplier Abcam Supplier No. ab109800250ug
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed for quantitative immunocapture of Complex III, cited in over 5 publications.

  • Species reactivity: Mouse, Rat, Cow, Human
  • Results type: Quantitative
  • Citation count: Cited in > 5 publications

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Product Type Western Blot Kit
Format Kit
Target Molecule Complex III
Cross Reactivity Mouse, Rat, Cow, Human
Quantity 250 μg
For Use With (Application) Immunoprecipitation
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.