missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement Factor H Antibody (L20/3) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
222.00€ - 531.00€
Specifications
| Antigen | Complement Factor H |
|---|---|
| Clone | L20/3 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:100-1:2000, ELISA 1:16000 |
| Applications | Western Blot, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18196869
|
Novus Biologicals
NBP1-05125 |
100 μg |
531.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18281243
|
Novus Biologicals
NBP1-05125-0.025MG |
0.025 mg |
222.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Complement Factor H Monoclonal specifically detects Complement Factor H in Human samples. It is validated for Western Blot, ELISA.Specifications
| Complement Factor H | |
| 1.0 mg/mL | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| 3075 | |
| complement factor H in human serum and plasma (1) | |
| Primary | |
| Store at 4C. Do not freeze. |
| L20/3 | |
| Western Blot 1:100-1:2000, ELISA 1:16000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| adrenomedullin binding protein, age-related maculopathy susceptibility 1, AHUS1, AMBP1, ARMD4, beta-1H, beta-1-H-globulin, CFHL3, complement factor H, factor H, factor H-like 1, FH, FHL1, H factor 1, H factor 1 (complement), H factor 2 (complement), HF, HF1ARMS1, HF2, HUSMGC88246 | |
| CFH | |
| IgG1 κ | |
| Protein A or G purified | |
| Specific for complement factor H in human serum and plasma (1). |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title