missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement Factor D/Adipsin Antibody (S09-6A9), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19629-50ul
This item is not returnable.
View return policy
Description
Complement Factor D/Adipsin Monoclonal antibody specifically detects Complement Factor D/Adipsin in Human samples. It is validated for Western Blot
Specifications
| Complement Factor D/Adipsin | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| S09-6A9 | |
| Unconjugated | |
| Adipsin, ADNcomplement factor D, C3 convertase activator, complement factor D (adipsin), complement factor D preproprotein, D component of complement (adipsin), DF, EC 3.4.21, EC 3.4.21.46, PFD, Properdin factor DADIPSIN | |
| Recombinant protein of human Complement Factor D/Adipsin (Uniprot # P00746) | |
| 50 μg | |
| Immunology | |
| 1675 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction