missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement C6 Antibody (056B-214.2.4.2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64337
This item is not returnable.
View return policy
Description
Complement C6 Monoclonal specifically detects Complement C6 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
| Complement C6 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| complement component 6, complement component C6 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| NB100-64337 recognizes complement component 6 (C6), a 104 kDa member of the complement C6/C7/C8/C9 family present in blood serum. The factor I modules of C6 bind the C-terminus of the C5b alpha-chain, anchoring the molecule to the membrane; they also binds complement component 7. This enables the formation of the C5b-7 precursor complex. C5b-7 then functions as a receptor for components C8 and C9 which form the complement membrane attack complex (MAC). Deficiency of C6 results in susceptibility to bacterial infection. NB100-64337 recognizes MAC. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| 056B-214.2.4.2 | |
| Unconjugated | |
| P13671 | |
| C6 | |
| Purified human C6 | |
| 0.05 mg | |
| Immunology, Innate Immunity | |
| 729 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction