missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement C3d Antibody (053A-1149.3.1.4) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
785.00€
Specifications
| Antigen | Complement C3d |
|---|---|
| Clone | 053A-1149.3.1.4 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
| Applications | Western Blot |
Description
Complement C3d Monoclonal specifically detects Complement C3d in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| Complement C3d | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Human | |
| P01024 | |
| 718 | |
| Purified C3d from human plasma | |
| Primary | |
| NB100-65510 is specific for human complement component 3d (C3d), a 33kDa polypeptide fragment generated over the course of complement activation where complement 3 (C3) convertases cleave C3 to C3b, which is further degraded into iC3b and C3dg/C3d. C3d is involved in the regulation of many aspects of the immune response, including antigen processing and presentation. The C3d/antigen complex mediates B cell activation by simultaneously binding antigen-specific surface bound immunoglobulin and CD21, lowering the complement activation threshold. C3 is crucial in the induction of tolerance generated when an antigen is introduced into immunoprivileged sites and this is exploited by pathogens and cancer cells to evade the immune system by inhibiting complement activation. |
| 053A-1149.3.1.4 | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| Monoclonal | |
| Mouse | |
| BBS (pH 8.4) with 0.1% sodium azide | |
| AHUS5, ARMD9, ASP, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1, complement C3, complement component 3, complement component C3, CPAMD1acylation-stimulating protein cleavage product | |
| C3 | |
| IgG1 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title