Learn More
Cytiva Column Holder
Product Code. 11300312 Shop All Cytiva Products
Change view
Click to view available options
Packaging Quantity:
1 Each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging Quantity Unit Size
11300312 1 Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11300312 Supplier Cytiva Supplier No. 28956282
Estimated Shipment: 25-11-2026
Holds ÅKTA columns. Cytiva Column Holder for columns with outer diameter between 10 and 50mm.
  • Attached to ÅKTA avant or ÅKTA pure using the rail system
TRUSTED_SUSTAINABILITY
Specifications
Type Column Holder
Packaging Quantity 1 Each
For Use With ÄKTA Avant or ÄKTA Pure Using The Rail System