missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Collagen X alpha 1 Antibody (SR3302), Novus Biologicals™

Rabbit Monoclonal Antibody

327.00€ - 382.00€

Specifications

Antigen Collagen X alpha 1
Clone SR3302
Dilution Western Blot 1:500-1:2000
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30041756
View Documents
Novus Biologicals™
NBP3-27767-100ul
100 μL
382.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30041784
View Documents
Novus Biologicals™
NBP3-27767-50ul
50 μL
327.00€
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Collagen X alpha 1 Monoclonal antibody specifically detects Collagen X alpha 1 in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications

Specifications

Collagen X alpha 1
Western Blot 1:500-1:2000
Monoclonal
Purified
RUO
Human, Mouse, Rat
COL10A1, Collagen Alpha-1(X) Chain, Collagen X, Alpha-1 Polypeptide, Schmid Metaphyseal Chondrodysplasia
A synthesized peptide derived from human Collagen X alpha 1 (Uniprot #: Q03692)
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
SR3302
Western Blot
Unconjugated
Rabbit
Cell Biology, Cytoskeleton Markers, Signal Transduction
PBS (pH 7.4), 150mM NaCl, 50% glycerol
1300
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.