missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen VII Mouse anti-Human, Bovine, Equine, Guinea Pig, Goat, Sheep, Clone: LH7.2, Novus Biologicals™
Mouse Monoclonal Antibody
519.00€
Specifications
| Antigen | Collagen VII |
|---|---|
| Clone | LH7.2 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1 to 2 μg/mL, Flow Cytometry 1 to 2 μg/million cells, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence |
Description
Collagen VII Monoclonal specifically detects Collagen VII in Human, Bovine, Equine, Guinea Pig, Goat, Sheep samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Collagen VII | |
| 0.2 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Cell Biology, Cellular Markers, Extracellular Matrix | |
| 10 mM PBS with 0.05% BSA | |
| 1294 | |
| IgG1 κ | |
| Protein A or G purified |
| LH7.2 | |
| Western Blot 1 to 2 μg/mL, Flow Cytometry 1 to 2 μg/million cells, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Bovine, Equine, Guinea Pig, Goat | |
| Collagen 7, collagen alpha-1(VII) chain, collagen VII, alpha-1 polypeptide, collagen, type VII, alpha 1, Collagen-7, EBD1, EBDCT, EBR1, epidermolysis bullosa, dystrophic, dominant and recessive, LC collagen, long chain collagen, Long-chain collagen | |
| Recombinant full-length human Collagen VII protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title