missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen IV Rabbit anti-Human, Clone: COL4/4241R, Novus Biologicals™
Rabbit Monoclonal Antibody
309.00€
Specifications
| Antigen | Collagen IV |
|---|---|
| Clone | COL4/4241R |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml |
| Applications | Immunohistochemistry (Paraffin) |
Description
Collagen IV Monoclonal specifically detects Collagen IV in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
| Collagen IV | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cellular Markers, Extracellular Matrix, Stem Cell Markers | |
| 10 mM PBS with 0.05% BSA | |
| Purified human placental extract | |
| Primary | |
| Store at 4°C. |
| COL4/4241R | |
| Immunohistochemistry-Paraffin 1-2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 1282 | |
| IgG | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title