missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen IV alpha 1 Antibody (C3), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-43263
This item is not returnable.
View return policy
Description
Collagen IV alpha 1 Monoclonal antibody specifically detects Collagen IV alpha 1 in Human samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| Collagen IV alpha 1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 0.2-2 μg/mL, Immunohistochemistry 5-20 μg/mL, Immunocytochemistry/ Immunofluorescence 5-20 μg/mL, Immunoprecipitation | |
| alpha-1 chain, arresten, COL4A1 NC1 domain, collagen alpha-1(IV) chain, collagen IV, alpha-1 polypeptide, collagen, type IV, alpha 1, Collagen-4, EC 3.4.23, EC 6.3.1.2 | |
| Immunized with recombinant Human Collagen IV alpha 1 (Uniprot#: P02462; His1444∽Thr1669) | |
| 100 μL | |
| Cell Biology, Cellular Markers, Extracellular Matrix, Stem Cell Markers | |
| 1282 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence | |
| C3 | |
| Unconjugated | |
| PBS, pH 7.4, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction