Learn More
COL4A3 Antibody (COL4A3/9395R), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-44590-100ug
427.75 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
COL4A3 Monoclonal antibody specifically detects COL4A3 in Human samples. It is validated for Immunohistochemistry
Specifications
| COL4A3 | |
| Monoclonal | |
| 0.2 mg/ml | |
| 10 mM PBS with 0.05% BSA | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| 1285 | |
| Store at 4⪚C. | |
| IgG κ |
| Immunohistochemistry | |
| COL4A3/9395R | |
| Unconjugated | |
| ATS2, ATS3, collagen alpha-3(IV) chain, collagen IV, alpha-3 polypeptide, collagen, type IV, alpha 3 (Goodpasture antigen), tumstatin | |
| Recombinant fragment (around aa1400-1670) of human COL4A3 protein (Uniprot: P02462, P08572, P29400, P53420). Exact sequence is proprietary) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |