missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDNRA/Endothelin R Type A Antibody, Novus Biologicals™
Sheep Polyclonal Antibody has been used in 4 publications
Brand: Novus Biologicals NB600-836
This item is not returnable.
View return policy
Description
EDNRA/Endothelin R Type A Polyclonal specifically detects EDNRA/Endothelin R Type A in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| EDNRA/Endothelin R Type A | |
| Polyclonal | |
| Unconjugated | |
| PBS with No Preservative | |
| EDNRA | |
| Synthetic peptide corresponding to amino acids 410-422 (QEQNHNTERSSHK) of Rat Endothelin A Receptor, conjugated to KLH. | |
| 0.125 mg | |
| GPCR, Neuroscience, Signal Transduction | |
| 1909 | |
| Mouse | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 1.7 mg/mL | |
| Western Blot 1:100-1:2000, Immunohistochemistry 10-20 ug/ml, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 20-40 ug/ml, Immunohistochemistry-Paraffin 1:10-1:500 | |
| endothelin receptor type A, endothelin-1 receptor, endothelin-1-specific receptor, ET1-specific type, ETAR, ETRA, G protein-coupled receptor, hET-AR | |
| Sheep | |
| Protein G purified | |
| RUO | |
| Primary | |
| This recognizes epitopes on the cytoplasmic side of the receptor and will not block the binding of endothelins. Therefore, the cell membrane must be permeable to the. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction