missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COL13A1 Monoclonal Antibody (584718)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561655
This item is not returnable.
View return policy
Description
Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.
This gene encodes the alpha chain of one of the nonfibrillar collagens. The function of this gene product is not known, however, it has been detected at low levels in all connective tissue-producing cells so it may serve a general function in connective tissues. Unlike most of the collagens, which are secreted into the extracellular matrix, collagen XIII contains a transmembrane domain and the protein has been localized to the plasma membrane. The transcripts for this gene undergo complex and extensive splicing involving at least eight exons. Like other collagens, collagen XIII is a trimer; it is not known whether this trimer is composed of one or more than one alpha chain isomer. A number of alternatively spliced transcript variants have been described, but the full length nature of some of them has not been determined. [provided by RefSeq, Jul 2008].
Specifications
| COL13A1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q5TAT6 | |
| COL13A1 | |
| Chinese hamster ovary cell line CHO-derived recombinant human Collagen XIII alpha 1, Glu109-Pro668. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin) | |
| 584718 | |
| Unconjugated | |
| COL13A1 | |
| CMS19; COL13A1; Collagen alpha-1(XIII) chain; collagen type XIII alpha 1 chain; collagen, type XIII, alpha 1; COLXIIIA1 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 1305 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction