missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Coilin Antibody (Pdelta) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB600-1275-0.025ml
This item is not returnable.
View return policy
Description
Coilin Monoclonal specifically detects Coilin in Human, Mouse (Negative) samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Microarray.
Specifications
| Coilin | |
| Monoclonal | |
| 1.5 mg/mL | |
| Western Blot 1-2 μg/mL, Immunocytochemistry/Immunofluorescence 10 μg/mL, Microarray | |
| CLN80p80-coilin, coilin, coilin p80, p80 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Coilin (Pdelta) | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
| Western Blot | |
| Pdelta | |
| Unconjugated | |
| P38432 | |
| COIL | |
| Recombinant fragment (C-terminus of Human coilin, 389 amino acids). | |
| 0.025 mL | |
| Neuroscience | |
| 8161 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction