Learn More
CNPY2 Antibody (8F6), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-42945
This item is not returnable.
View return policy
Description
CNPY2 Monoclonal antibody specifically detects CNPY2 in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| CNPY2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| 8F6 | |
| Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry 1:50 - 1:500, Immunocytochemistry/ Immunofluorescence 1:50 - 1:500 | |
| canopy 2 homolog (zebrafish), HP10390, MSAPprotein canopy homolog 2, Putative secreted protein Zsig9, TMEM4CNPY4, Transmembrane protein 4MIR-interacting saposin-like protein, ZSIG9Cnpy2 | |
| canopy 2 homolog(zebrafish) - (Uniprot# Q9Y2B0) | |
| 100 μg | |
| Cell Biology | |
| 10330 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG2a |