missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNPase Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
224.00€ - 471.00€
Specifications
| Antigen | CNPase |
|---|---|
| Applications | Western Blot, Immunohistochemistry, KnockDown |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18393087
|
Novus Biologicals
NBP3-03492-20UL |
20 μL |
224.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18322324
|
Novus Biologicals
NBP3-03492-100UL |
100 μL |
471.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CNPase Polyclonal specifically detects CNPase in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockout Validated.Specifications
| CNPase | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| 2', 3' cyclic nucleotide 3' phosphohydrolase, 2'-3'-cyclic nucleotide 3' phosphodiesterase, 2'-3'-cyclic-nucleotide 3'-phosphodiesterase, CNP1, CNPase, EC 3.1.4.37 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 152-421 of human CNPase (NP_149124.3). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, KnockDown | |
| Unconjugated | |
| Rabbit | |
| Cellular Markers, Glia Markers, Neuronal Cell Markers, Neuroscience, Oligodendrocyte Cell Markers | |
| PBS with 50% glycerol, pH7.3. | |
| 1267 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title