Learn More
CNKSR3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14084-1-AP-20UL
This item is not returnable.
View return policy
Description
CNKSR3 may be involved in transepithelial sodium transport. Regulates aldosterone-induced and ENaC-mediated sodium transport possibly through regulation of the ERK pathway.Specifications
| CNKSR3 | |
| Polyclonal | |
| Unconjugated | |
| CNKSR3 | |
| CNK homolog protein 3, CNK3, CNKSR family member 3, CNKSR3, Connector enhancer of KSR 3, MAGI1, Maguin like protein | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 154043, 215748, 308113 | |
| -20°C | |
| Liquid |
| Western Blot, Immunoprecipitation | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5SGD7, Q6P9H4, Q8BMA3 | |
| CNKSR3 | |
| CNKSR3 Fusion Protein Ag5213 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |