Learn More
IBA Lifesciences CMV IE-1 peptide QIKVRVDMV (HLA-B*0801)
GREENER_CHOICE
Product Code. 15392146 Shop All IBA Lifesciences Products
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Product Code. 15392146
Brand: IBA Lifesciences 67017901
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Single peptide synthesized as presented by the MHC class I allele
  • CMV IE-1 peptide QIKVRVDMV (HLA-B*0801) is a single peptide for stimulation of T cells.
  • The peptide from human cytomegalovirus (CMV) Immediate-Early (IE) is synthesized as it is presented by the MHC class I HLA-B*0801 allele and can be used for targeted in vitro generation and expansion of antigen-specific CD8+ T cells.
  • The peptides were purified by HPLC using a water and acetonitrile gradient and are endotoxin-free.
  • Specifications
    Content And Storage Recommended: -25°C to -15°C
    Quantity 1 mg