Learn More
CLP Contract Required Detection Limit (CRDL) Solution 1: Pb @ 6; Be, Cd, Se @ 10; Ag, As, Cr, Tl @ 20; Mn @ 30; Zn @ 40; Cu @ 50; Ni @ 80; Co, V @ 100; Sb @ 120 μg/mL in 5% HNO3, tr. Tartaric Acid, VHG™
Product Code. 30003139
Change view
Click to view available options
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
30003139 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 30003139 Supplier VHG Labs Supplier No. VHGCRDL100
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
CLP Contract Required Detection Limit (CRDL) Solution 1: Pb @ 6; Be, Cd, Se @ 10; Ag, As, Cr, Tl @ 20; Mn @ 30; Zn @ 40; Cu @ 50; Ni @ 80; Co, V @ 100; Sb @ 120 μg/mL in 5% HNO3, tr. Tartaric Acid
TRUSTED_SUSTAINABILITY
Specifications
Solvent or Matrix Dilute HNO3
Quantity 100 mL