Learn More
Clomifene Citrate, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Clomifene Citrate
Specifications
Specifications
| Chemical Name or Material | Clomiphene Citrate |
| CAS | 50-41-9 |
| Analyte or Component Names | Clomiphene Citrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C26 H28 Cl N O . C6 H8 O7 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Clomifene Citrate, Ethanamine, 2-[4-(2-chloro-1,2-diphenylethenyl)phenoxy]-N,N-diethyl-, 2-hydroxy-1,2,3-propanetricarboxylate (1:1), Triethylamine, 2-[p-(2-chloro-1,2-diphenylvinyl)phenoxy]-, citrate (1:1) (8CI), Triethylamine, 2-[p-(2-chloro-1,2-diphenylvinyl)phenoxy]-, citrate (6CI,7CI), 2-[p-(2-Chloro-1,2-diphenylvinyl)phenoxy]triethylamine dihydrogen citrate, Chloramiphene, Clomid, Clomifeno, Clomiphene citrate, Clomiphene dihydrogen citrate, Clomivid, Clomphid, Clostilbegit, Clostilbegyt, Dyneric, Fertivet, Fertyl, Genozym, Ikaclomin, Ikaclomine, MRL 41, Mer 41, NSC 35770, Omifin, Pergotime, Racemic clomiphene citrate, Serophene, 1-[p-(beta-Diethylaminoethoxy)phenyl]-1,2-diphenyl-2-chloroethylene citrate |
| InChI Formula | InChI=1S/C26H28ClNO.C6H8O7/c1-3-28(4-2)19-20-29-24-17-15-22(16-18-24)25(21-11-7-5-8-12-21)26(27)23-13-9-6-10-14-23;7-3(8)1-6(13,5(11)12)2-4(9)10/h5-18H,3-4,19-20H2,1-2H3;13H,1-2H2,(H,7,8)(H,9,10)(H,11,12) |
| SMILES | CCN(CC)CCOc1ccc(cc1)C(=C(Cl)c2ccccc2)c3ccccc3.OC(=O)CC(O)(CC(=O)O)C(=O)O |
| IUPAC Name | 2-[4-(2-chloro-1,2-diphenylethenyl)phenoxy]-N,N-diethylethanamine;2-hydroxypropane-1,2,3-tricarboxylic acid |
| Show More |