missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Clomifene Citrate, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Clomiphene Citrate |
| CAS | 50-41-9 |
| Analyte or Component Names | Clomiphene Citrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C26 H28 Cl N O . C6 H8 O7 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Clomifene Citrate, Ethanamine, 2-[4-(2-chloro-1,2-diphenylethenyl)phenoxy]-N,N-diethyl-, 2-hydroxy-1,2,3-propanetricarboxylate (1:1), Triethylamine, 2-[p-(2-chloro-1,2-diphenylvinyl)phenoxy]-, citrate (1:1) (8CI), Triethylamine, 2-[p-(2-chloro-1,2-diphenylvinyl)phenoxy]-, citrate (6CI,7CI), 2-[p-(2-Chloro-1,2-diphenylvinyl)phenoxy]triethylamine dihydrogen citrate, Chloramiphene, Clomid, Clomifeno, Clomiphene citrate, Clomiphene dihydrogen citrate, Clomivid, Clomphid, Clostilbegit, Clostilbegyt, Dyneric, Fertivet, Fertyl, Genozym, Ikaclomin, Ikaclomine, MRL 41, Mer 41, NSC 35770, Omifin, Pergotime, Racemic clomiphene citrate, Serophene, 1-[p-(beta-Diethylaminoethoxy)phenyl]-1,2-diphenyl-2-chloroethylene citrate |
| InChI Formula | InChI=1S/C26H28ClNO.C6H8O7/c1-3-28(4-2)19-20-29-24-17-15-22(16-18-24)25(21-11-7-5-8-12-21)26(27)23-13-9-6-10-14-23;7-3(8)1-6(13,5(11)12)2-4(9)10/h5-18H,3-4,19-20H2,1-2H3;13H,1-2H2,(H,7,8)(H,9,10)(H,11,12) |
| Solution Type | Neat |
| SMILES | CCN(CC)CCOc1ccc(cc1)C(=C(Cl)c2ccccc2)c3ccccc3.OC(=O)CC(O)(CC(=O)O)C(=O)O |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction