Learn More
Clofibrate, 95+%, Thermo Scientific™
18.00€ - 236.00€
Chemical Identifiers
| CAS | 637-07-0 |
|---|---|
| Molecular Formula | C12H15ClO3 |
| Molecular Weight (g/mol) | 242.70 |
| MDL Number | MFCD00000615 |
| InChI Key | KNHUKKLJHYUCFP-UHFFFAOYSA-N |
| Synonym | clofibrate, ethyl clofibrate, clofibratum, clofibrato, atromid-s, miscleron, atromid, liprin, epib, angiokapsul |
| PubChem CID | 2796 |
| ChEBI | CHEBI:3750 |
| IUPAC Name | ethyl 2-(4-chlorophenoxy)-2-methylpropanoate |
| SMILES | CCOC(=O)C(C)(C)OC1=CC=C(Cl)C=C1 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15483119
|
Thermo Scientific Alfa Aesar
J60342.03 |
1g |
18.00€
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15493119
|
Thermo Scientific Alfa Aesar
J60342.06 |
5g |
29.70€
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15403129
|
Thermo Scientific Alfa Aesar
J60342.14 |
25g |
86.40€
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15413129
|
Thermo Scientific Alfa Aesar
J60342.22 |
100g |
236.00€
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
SolubilitySoluble to 100 mM in DMSO.
Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents, heat, bases. Store in refrigerator.
Chemical Identifiers
| C12H15ClO3 | |
| MFCD00000615 | |
| clofibrate, ethyl clofibrate, clofibratum, clofibrato, atromid-s, miscleron, atromid, liprin, epib, angiokapsul | |
| CHEBI:3750 | |
| CCOC(=O)C(C)(C)OC1=CC=C(Cl)C=C1 |
Specifications
| 637-07-0 | |
| 154°C to 156°C | |
| C12H15ClO3 | |
| MFCD00000615 | |
| 1913459 | |
| clofibrate, ethyl clofibrate, clofibratum, clofibrato, atromid-s, miscleron, atromid, liprin, epib, angiokapsul | |
| KNHUKKLJHYUCFP-UHFFFAOYSA-N | |
| ethyl 2-(4-chlorophenoxy)-2-methylpropanoate | |
| 2796 | |
| 242.7 | |
| Clofibrate |
| 1.14 | |
| 113°C (235°F) | |
| 1.5 | |
| UN3082 | |
| 14,2377 | |
| Soluble to 100 mM in DMSO. | |
| CCOC(=O)C(C)(C)OC1=CC=C(Cl)C=C1 | |
| 242.70 | |
| CHEBI:3750 | |
| ≥95% |
Safety and Handling
GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
H302-H315-H318-H335
missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
missing translation for 'einecsNumber' : 211-277-4
missing translation for 'rtecsNumber' : UE9480000
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.