missing translation for 'onlineSavingsMsg'
Learn More

Clofibrate, 95+%, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar J60342.03

Quantity 1g

 View more versions of this product

Product Code. 15483119

  • 18.00€ / 1g

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Soluble to 100 mM in DMSO.

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents, heat, bases. Store in refrigerator.
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

637-07-0
242.70
KNHUKKLJHYUCFP-UHFFFAOYSA-N
2796
ethyl 2-(4-chlorophenoxy)-2-methylpropanoate
C12H15ClO3
MFCD00000615
clofibrate, ethyl clofibrate, clofibratum, clofibrato, atromid-s, miscleron, atromid, liprin, epib, angiokapsul
CHEBI:3750
CCOC(=O)C(C)(C)OC1=CC=C(Cl)C=C1

Specifications

637-07-0
154°C to 156°C
C12H15ClO3
MFCD00000615
UN3082
14,2377
Soluble to 100 mM in DMSO.
CCOC(=O)C(C)(C)OC1=CC=C(Cl)C=C1
242.70
CHEBI:3750
≥95%
1.14
113°C (235°F)
1.5
1g
1913459
clofibrate, ethyl clofibrate, clofibratum, clofibrato, atromid-s, miscleron, atromid, liprin, epib, angiokapsul
KNHUKKLJHYUCFP-UHFFFAOYSA-N
ethyl 2-(4-chlorophenoxy)-2-methylpropanoate
2796
242.7
Clofibrate
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.